missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-fluorophényl)phosphine, 98+%

Numéro de catalogue. AC306340010
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
Packaging:
Bouteille en verre
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC306340010 1 g Bouteille en verre
AC306340050 5 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC306340010 Fournisseur Thermo Scientific Chemicals Code fournisseur 306340010
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

CAS : 18437-78-0 | C18H12F3P | 316,26 g/mol

Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

Identifiants chimiques

CAS 18437-78-0
Molecular Formula C18H12F3P
Molecular Weight (g/mol) 316.26
MDL Number MFCD00013553
InChI Key GEPJPYNDFSOARB-UHFFFAOYSA-N
Synonym tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi
PubChem CID 140387
IUPAC Name tris(4-fluorophényl)phosphane
SMILES FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1

Spécifications

Melting Point 79.0°C to 82.0°C
Color Du blanc au jaune
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Bouteille en verre
Linear Formula (FC6H4)3P
Quantity 1 g
Formula Weight 316.27
Percent Purity 98+%
Physical Form Poudre cristalline
Chemical Name or Material Tris(4-fluorophenyl)phosphine
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Utilisation uniquement de la recherche RUO –