missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-fluorophenyl)phosphine, 98%

Catalog No. AAA1481314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1481314 25 g
AAA1481303 1 g
AAA1481306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1481314 Supplier Thermo Scientific Chemicals Supplier No. A1481314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18437-78-0 | C18H12F3P | 316.26 g/mol

Tris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.Stable under recommended storage conditions.

Chemical Identifiers

CAS 18437-78-0
Molecular Formula C18H12F3P
Molecular Weight (g/mol) 316.26
MDL Number MFCD00013553
InChI Key GEPJPYNDFSOARB-UHFFFAOYSA-N
Synonym tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi
PubChem CID 140387
IUPAC Name tris(4-fluorophenyl)phosphane
SMILES FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1

Specifications

Melting Point 78°C to 82°C
Quantity 25 g
Beilstein 919838
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 316.27
Percent Purity 98%
Chemical Name or Material Tris(4-fluorophenyl)phosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only