missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(4-aminophényl)méthane 97,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 208°C |
| Quantity | 5 g |
| Formula Weight | 289.38 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Tris(4-aminophenyl)methane |
Sécurité et manipulation
| TSCA | No |