missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Tris(3-Hydroxypropyltriazolylmethyl)Amine
Description
Tris(3-Hydroxypropyltriazolylmethyl)Amine
Spécifications
Spécifications
| CAS | 760952-88-3 |
| Melting Point | 331°C to 336°C |
| Molecular Formula | C18H30N10O3 |
| Linear Formula | C18H30N10O3 |
| MDL Number | MFCD27665386 |
| Quantity | 1 g |
| Synonym | 3,3',3''-(4,4',4''-(Nitrilotris(methylene))tris(1H-1,2,3-triazole-4,1-diyl))tris(propan-1-ol); THPTA |
| Molecular Weight (g/mol) | 434.5 g/mol |