missing translation for 'onlineSavingsMsg'
Learn More

Tris(3,5-dimethyl-1-pyrazolyl)methane 98.0+%, TCI America™

Catalog No. T3025200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3025200MG 200 mg
T30251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3025200MG Supplier TCI America Supplier No. T3025200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28791-97-1
Molecular Formula C16H22N6
Molecular Weight (g/mol) 298.39
MDL Number MFCD00958749
InChI Key COKUMKYWINNWLV-UHFFFAOYSA-N
PubChem CID 4359225
IUPAC Name 1-[bis(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-3,5-dimethyl-1H-pyrazole
SMILES CC1=NN(C(N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 200 mg
Formula Weight 298.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(3,5-dimethyl-1-pyrazolyl)methane