missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methoxyphenyl)bismuthine 97.0+%, TCI America™

Catalog No. T18381G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18381G Supplier TCI America Supplier No. T18381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83724-41-8
Molecular Formula C21H21BiO3
Molecular Weight (g/mol) 530.376
MDL Number MFCD00092705
InChI Key VFWRGMGLLNCHIA-UHFFFAOYSA-N
Synonym tris 2-methoxyphenyl bismuthine, tris 2-methoxyphenyl bismuthane, acmc-209pt3, tris-2-methoxyphenyl bismuthane, bismuthine,tris 2-methoxyphenyl
PubChem CID 2734045
IUPAC Name tris(2-methoxyphenyl)bismuthane
SMILES COC1=CC=CC=C1[Bi](C2=CC=CC=C2OC)C3=CC=CC=C3OC

Specifications

Melting Point 163°C
Color White
Quantity 1 g
Formula Weight 530.38
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-methoxyphenyl)bismuthine