missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-ethylhexyl) Trimellitate 97.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 414°C |
| Quantity | 500 mL |
| Formula Weight | 546.79 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tris(2-ethylhexyl) Trimellitate |
Sécurité et manipulation
| EINECSNumber | (3)-1372&(3)-2684 |
| RTECSNumber | DC2080000 |
| TSCA | Yes |