missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-ethylhexyl) Phosphite 90.0+%, TCI America™

Catalog No. P1169500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1169500ML 500 mL
P116925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1169500ML Supplier TCI America Supplier No. P1169500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 301-13-3
Molecular Formula C24H51O3P
Molecular Weight (g/mol) 418.643
MDL Number MFCD00015266
InChI Key ILLOBGFGKYTZRO-UHFFFAOYSA-N
Synonym Phosphorous Acid Trioctyl Ester, Phosphorous Acid Tris(2-ethylhexyl) Ester, Trioctyl Phosphite
PubChem CID 9320
IUPAC Name tris(2-ethylhexyl) phosphite
SMILES CCCCC(CC)COP(OCC(CC)CCCC)OCC(CC)CCCC

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 418.64
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-ethylhexyl) Phosphite
RTECSNumber TH2090000
TSCA Yes