missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-ethylhexyl)amine 93.0+%, TCI America™

Catalog No. T117825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T117825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T117825ML Supplier TCI America Supplier No. T117825ML
Only null left

Chemical Identifiers

CAS 1860-26-0
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.679
MDL Number MFCD00042903
InChI Key BZUDVELGTZDOIG-UHFFFAOYSA-N
Synonym Trioctylamine
PubChem CID 102705
IUPAC Name 2-ethyl-N,N-bis(2-ethylhexyl)hexan-1-amine
SMILES CCCCC(CC)CN(CC(CC)CCCC)CC(CC)CCCC

Specifications

Color Yellow
Quantity 25 mL
UN Number 2735
Formula Weight 353.68
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-ethylhexyl)amine
EINECSNumber (2)-0143
TSCA Yes