missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-butoxyethyl) phosphate, 95%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Density | 1.0000g/mL |
| Boiling Point | 215°C to 228°C (4.0mmHg) |
| Flash Point | 132°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4370 to 1.439 |
| Linear Formula | [CH3(CH2)3OCH2CH2O]3P(O) |
| Quantity | 500g |
| Specific Gravity | 1 |
| Show More |
Safety and Handling
| EINECSNumber | 201-122-9 |
RUO – Research Use Only