missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-aminoethyl)amine, 98+%

Catalog No. AC314791000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314791000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC314791000 Supplier Thermo Scientific Chemicals Supplier No. 314791000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4097-89-6 | C6H18N4 | 146.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4097-89-6
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.24
InChI Key MBYLVOKEDDQJDY-UHFFFAOYSA-N
Synonym tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
PubChem CID 77731
ChEBI CHEBI:30631
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
SMILES C(CN(CCN)CCN)N

Specifications

Melting Point -16.0°C
Color Yellow
Density 0.9770g/mL
Boiling Point 265.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980
Linear Formula (H2NCH2CH2)3N
Quantity 100 mL
Beilstein 04,II,695
Specific Gravity 0.977
Solubility Information Solubility in water: soluble.
Formula Weight 146.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tris(2-aminoethyl)amine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
EINECSNumber 223-857-4

RUO – Research Use Only