missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,3-dibromopropyl) Isocyanurate 97.0+%, TCI America™

Catalog No. I0355500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0355500G 500 g
I035525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0355500G Supplier TCI America Supplier No. I0355500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52434-90-9
Molecular Formula C12H15Br6N3O3
Molecular Weight (g/mol) 728.694
MDL Number MFCD00010290
InChI Key NZUPFZNVGSWLQC-UHFFFAOYSA-N
Synonym tris 2,3-dibromopropyl isocyanurate, 1,3,5-tris 2,3-dibromopropyl-1,3,5-triazinane-2,4,6-trione, hexahydro-1,3,5-tris 2,3-dibromopropyl-1,3,5-triazine-2,4,6-trione, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3,5-tris 2,3-dibromopropyl, 1,3,5-tris 2,3-dibromopropyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione, isocyanuric acid tris 2,3-dibromopropyl ester, 1,3,5-tris 2,3-dibromopropyl-s-triazine-2,4,6-trione, acmc-1apwt, tri 2,3-dibromopropyl isocyanate
PubChem CID 103634
IUPAC Name 1,3,5-tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione
SMILES C(C(CBr)Br)N1C(=O)N(C(=O)N(C1=O)CC(CBr)Br)CC(CBr)Br

Specifications

Melting Point 110°C
Color White
Quantity 500 g
Formula Weight 728.69
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,3-dibromopropyl) Isocyanurate
EINECSNumber (5)-1049
TSCA No