missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, 98+%

Catalog No. AA1278304
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1278304 2 g
AA1278302 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1278304 Supplier Thermo Scientific Chemicals Supplier No. 01278304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50525-27-4 | C30H36Cl2N6O6Ru | 748.62 g/mol

Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate is a catalyst used in light-induced redox reactions. It is an important catalyst in studies towards photochemical water splitting. This product can be used as a catalyst in pharmaceutical research. It is widely applied in fields of photoelectrical chemical components, photovoltaic cells, smart biosensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50525-27-4
Molecular Formula C30H36Cl2N6O6Ru
Molecular Weight (g/mol) 748.62
MDL Number MFCD00149670
InChI Key WHELTKFSBJNBMQ-UHFFFAOYSA-L
Synonym tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
PubChem CID 131664161
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Melting Point 63°C to 66°C
Color Red to Orange
Assay Percent Range >98%
Quantity 2 g
Solubility Information Soluble in water.
Formula Weight 748.63 (640.54 Anhydrous)
Physical Form Crystalline
Chemical Name or Material Tris(2, 2'-bipyridine)dichlororuthenium(II) hexahydrate
Hazard Category H315-H319: Causes skin irritation. Causes serious eye irritation.
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
EINECSNumber 238-266-7
RTECSNumber VM2730000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only