missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Triptolide

Catalog No. 32531
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32531 1 mg
325310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32531 Supplier Tocris Bioscience™ Supplier No. 3253/1

Inhibits RNAPII-mediated transcription; antitumor, anti-inflammatory and immunosuppressive

Triptolide inhibits DNA-dependent ATPase activity of XBP and induces inhibition of RNA polymerase II (RNAPII)-mediated transcription (IC50 = 200 nM). Selective for RNAPII over RNAPI and RNAPIII. Triptolide blocks RNA synthesis in HeLa cells (IC50 = 62 nM); exhibits potent antiproliferative activity in 60 cancer cell lines (average IC50 = 12 nM) and induces apoptosis by blocking TNF-α-mediated c-IAP1 and c-IAP2 induction. Also displays immunosuppressive and anti-inflammatory activity.

Chemical Identifiers

CAS 38748-32-2
Molecular Formula C20H24O6

Specifications

Quantity 1 mg
Formula Weight 360.4
Purity >98%
Molecular Weight (g/mol) 360.41
Chemical Name or Material Triptolide
Recommended Storage Store at -20°C