missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triptolide

Catalog No. AC466840250
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466840250 25 mg
AC466841000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466840250 Supplier Thermo Scientific Chemicals Supplier No. 466840250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38748-32-2 | C20H24O6, C20H24O6 | 360.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38748-32-2
Molecular Formula C20H24O6

Specifications

Quantity 25 mg
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso, ethanol
Melting Point 226.0°C to 227.0°C
Formula Weight 360.41
MDL Number MFCD00210565
InChI Key DFBIRQPKNDILPW-CIVMWXNOSA-N
SMILES [H][C@@]12C[C@@H]3O[C@@]33[C@H](O)[C@]4(O[C@H]4[C@@H]4O[C@]34[C@@]1(C)CCC1=C2COC1=O)C(C)C
IUPAC Name (1S,2S,4S,5S,7R,8R,9S,11S,13S)-8-hydroxy-1-methyl-7-(propan-2-yl)-3,6,10,16-tetraoxaheptacyclo[11.7.0.0²,⁴.0²,⁹.0⁵,⁷.0⁹,¹¹.0¹⁴,¹⁸]icos-14(18)-en-17-one
Molecular Weight (g/mol) 360.41
Chemical Name or Material Triptolide
What storage conditions should I use for Triptolide?

We recommend storing Triptolide in powder form at -20°C, desiccated. Once in solution, it should be stored at -20°C and used within 1 month to prevent loss of potency. We suggest you make aliquots of the stock solution to avoid multiple freeze/thaw cycles.

How can I prepare a 10 mM stock solution of Triptolide?

To prepare a 10 mM stock of Triptolide, we recommend that you reconstitute 5 mg of powder in 1.38 mL of DMSO.

Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

RUO – Research Use Only