missing translation for 'onlineSavingsMsg'
Learn More

Tripropylene Glycol Monomethyl Ether (mixture of isomer) 93.0+%, TCI America™

Catalog No. T0651500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0651500G 500 g
T065125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0651500G Supplier TCI America Supplier No. T0651500G

Chemical Identifiers

CAS 25498-49-1
Molecular Formula C10H22O4
MDL Number MFCD00059142

Specifications

Melting Point -79°C
Color Yellow
Boiling Point 243°C
Quantity 500 g
Formula Weight 206.28
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripropylene Glycol Monomethyl Ether (mixture of isomer)
EINECSNumber (2)-0438&(7)-0097
RTECSNumber UB8070000
TSCA Yes