missing translation for 'onlineSavingsMsg'
Learn More

Tripropylamine, 98%

Catalog No. AC162690010
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162690010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162690010 Supplier Thermo Scientific Chemicals Supplier No. 162690010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102-69-2 | C9H21N | 143.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-69-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD00009363
InChI Key YFTHZRPMJXBUME-UHFFFAOYSA-N
Synonym tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
PubChem CID 7616
ChEBI CHEBI:38880
IUPAC Name N,N-dipropylpropan-1-amine
SMILES CCCN(CCC)CCC

Specifications

Melting Point -93.0°C
Color Colorless
Density 0.7600g/mL
Boiling Point 156.0°C
Flash Point 32.5°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. dipropylamine (GC)
Packaging Glass bottle
Refractive Index 1.4155 to 1.4175
Linear Formula (CH3CH2CH2)3N
Quantity 1 L
Beilstein 04, II, 623
Specific Gravity 0.76
Solubility Information Solubility in water: 2.6g/L (20°C). Other solubilities: soluble in most organic solvents
Viscosity 0.627 mPa.s (25°C)
Formula Weight 143.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tripropylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
May cause respiratory irritation.
Harmful to aquatic life wi
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several min
EINECSNumber 203-047-7

RUO – Research Use Only