missing translation for 'onlineSavingsMsg'
Learn More

TrippNT Pipet Stands

$296.00 - $369.86

Specifications

Color White
For Use With (Equipment) Most brands of repeating pipettors
Product Type Pipet Stand
View More Specs

Products 2
Catalog Number Mfr. No. Positions Size Price Quantity & Availability  
Catalog Number Mfr. No. Positions Size Price Quantity & Availability  
50856040
Encompass
View Documents Promotion Details
TrippNT
TRP 50071
3 Small
Each for $296.00
Only null left
 
50856041
Encompass
View Documents Promotion Details
TrippNT
TRP 50176
6 Large
Each for $369.86
Only null left
 
Description

Description

Protect pipettor tips by storing on these angled pipettor stands.

Specifications

Specifications

White
Most brands of repeating pipettors
Pipet Stand
Videos
SDS
Documents

Documents