missing translation for 'onlineSavingsMsg'
Learn More

TrippNT Pipet Stands

Catalog No. 50856040
Encompass
Change view
Click to view available options
Positions:
3
6
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions Size
50856040 3 Small
50856041 6 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50856040 Supplier TrippNT Supplier No. TRP 50071

Protect pipettor tips by storing on these angled pipettor stands.

Specifications

Color White
Positions 3
For Use With (Equipment) Most brands of repeating pipettors
Size Small
Product Type Pipet Stand