missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrippNT Pipet Stands
Description
Protect pipettor tips by storing on these angled pipettor stands.
Specifications
Specifications
| Color | White |
| Positions | 6 |
| For Use With (Equipment) | Most brands of repeating pipettors |
| Size | Large |
| Product Type | Pipet Stand |