missing translation for 'onlineSavingsMsg'
Learn More

TrippNT Maui Lab Island 2 Foot Lab Cart

Supplier:  TrippNT TRP 50591

Encompass

Catalog No. 50871533


Only null left

Description

Description

Specifications

Specifications

Mobile Lab Cabinet
24 x 24 x 35 in.
Lockable storage with easy-to-raise, space-saving roll top door; Four 3 inch non-marking casters, 2 with brakes; 1 pull-out shelf on drawer slides; two keys
Polyethylene, PVC
Small
4 x 3 in. non-marking casters, 2 with brakes
White
35 in.
24 in.
Polyethylene
72 lb.
24 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions