missing translation for 'onlineSavingsMsg'
Learn More

TrippNT Kimwipe Holder with Magnet Mount

Catalog No. NC0083426
Encompass
Change view
Click to view available options
Mounting:
Magnetic Mount
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
NC0083426 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC0083426 Supplier TrippNT Supplier No. TRP 50653

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color White
Mounting Magnetic Mount
For Use With (Application) Fits Kimberly Clark XL Delicate Lab Wiper Box 34155-S, or Shur-wipe 29712
Includes Magnet Mount