missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine 95.0+%, TCI America™

Catalog No. T051925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T051925G 25 g
T0519100G 100 g
T0519500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T051925G Supplier TCI America Supplier No. T051925G

Chemical Identifiers

CAS 603-35-0
Molecular Formula C18H15P
Molecular Weight (g/mol) 262.29
MDL Number MFCD00003043 MFCD20489348
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
Synonym triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin
PubChem CID 11776
IUPAC Name triphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 81°C
Color White
Boiling Point 260°C
Quantity 25 g
Formula Weight 262.29
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylphosphine
EINECSNumber (3)-2518
RTECSNumber SZ3500000
TSCA Yes