missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylphosphine Oxide 98.0+%, TCI America™
Supplier: TCI America T0625100G
Specifications
| Triphenylphosphine Oxide | |
| 156°C | |
| 360°C | |
| MFCD00002080 MFCD03458802 | |
| 3077 | |
| FIQMHBFVRAXMOP-UHFFFAOYSA-N | |
| (diphenylphosphoroso)benzene | |
| 13097 | |
| 278.29 | |
| Crystalline Powder |
| 791-28-6 | |
| White-Yellow | |
| C18H15OP | |
| 100 g | |
| triphenylphosphine oxide, phosphine oxide, triphenyl, triphenyl phosphorus oxide, triphenyl phosphine oxide, triphenylphosphane oxide, tppo, triphenylphosphineoxide, ph3po, diphenylphosphoroso benzene, triphenylphosphanoxid | |
| O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 278.29 | |
| CHEBI:36601 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 791-28-6 | |
| 278.29 | |
| FIQMHBFVRAXMOP-UHFFFAOYSA-N | |
| 13097 | |
| (diphenylphosphoroso)benzene |
| C18H15OP | |
| MFCD00002080 MFCD03458802 | |
| triphenylphosphine oxide, phosphine oxide, triphenyl, triphenyl phosphorus oxide, triphenyl phosphine oxide, triphenylphosphane oxide, tppo, triphenylphosphineoxide, ph3po, diphenylphosphoroso benzene, triphenylphosphanoxid | |
| CHEBI:36601 | |
| O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-2609
RTECSNumber : SZ1676000
TSCA : Yes