missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine Oxide 98.0+%, TCI America™

Catalog No. p-7136371
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T062525G 25 g
T0625100G 100 g
T0625500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T062525G Supplier TCI America Supplier No. T062525G
Only null left

Chemical Identifiers

CAS 791-28-6
Molecular Formula C18H15OP
Molecular Weight (g/mol) 278.29
MDL Number MFCD00002080 MFCD03458802
InChI Key FIQMHBFVRAXMOP-UHFFFAOYSA-N
Synonym triphenylphosphine oxide, phosphine oxide, triphenyl, triphenyl phosphorus oxide, triphenyl phosphine oxide, triphenylphosphane oxide, tppo, triphenylphosphineoxide, ph3po, diphenylphosphoroso benzene, triphenylphosphanoxid
PubChem CID 13097
ChEBI CHEBI:36601
IUPAC Name (diphenylphosphoroso)benzene
SMILES O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 156°C
Color White-Yellow
Boiling Point 360°C
Quantity 25 g
UN Number 3077
Formula Weight 278.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylphosphine Oxide
EINECSNumber (3)-2609
RTECSNumber SZ1676000
TSCA Yes