accessibility menu, dialog, popup

UserName

OP

CAS RN 791-28-6

IUPAC Name: (diphenylphosphoroso)benzene
Synonyms: triphenylphosphine oxide|triphenyl phosphine oxide|triphenylphosphanoxid|triphenyl phosphorus oxide
Molecular Weight (g/mol): 278.29
Molecular Formula: C18H15OP
InChi Key: FIQMHBFVRAXMOP-UHFFFAOYSA-N
SMILES: O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)