missing translation for 'onlineSavingsMsg'
Learn More

(Triphenylphosphine)gold(I) Chloride 95.0+%, TCI America™

Catalog No. T2994200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2994200MG 200 mg
T29941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2994200MG Supplier TCI America Supplier No. T2994200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14243-64-2
Molecular Formula C18H15AuClP
Molecular Weight (g/mol) 494.71
MDL Number MFCD00009588
InChI Key IFPWCRBNZXUWGC-UHFFFAOYSA-M
Synonym chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
PubChem CID 10874691
IUPAC Name chlorogold; triphenylphosphane
SMILES Cl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 247°C
Color White-Yellow
Quantity 200 mg
Formula Weight 494.71
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Triphenylphosphine)gold(I) Chloride