missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl Phosphate 99.0+%, TCI America™

Catalog No. P0272500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0272500G 500 g
P027225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0272500G Supplier TCI America Supplier No. P0272500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115-86-6
Molecular Formula C18H15O4P
Molecular Weight (g/mol) 326.29
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Synonym triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
PubChem CID 8289
ChEBI CHEBI:35033
IUPAC Name triphenyl phosphate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 50°C
Color White
Boiling Point 220°C
Quantity 500 g
UN Number 3077
Formula Weight 326.29
Percent Purity ≥99.0% (GC)
Physical Form Flakes
Chemical Name or Material Triphenyl Phosphate
EINECSNumber (3)-2522
RTECSNumber TC8400000
TSCA Yes