missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenyl Phosphate Solution, NMR Reference Standard, 3 mM in chloroform-d (99.8 atom % D), MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | NMR Tube Size: 5 mm 8 in. |
| Concentration | 3 mM in chloroform-d (99.8 atom% D) |
| Linear Formula | (C6H5O)3 PO |
| MDL Number | MFCD00003031 |
| Quantity | 0.7 mL |
| Formula Weight | 326.28 |
| Percent Purity | 99% (CP) |
| Grade | NMR Reference Standard |