Learn More
Thermo Scientific Chemicals Tripeptidyl Peptidase Inhibitor
Chemical Identifiers
| CAS | 184901-82-4 |
|---|---|
| Molecular Formula | C18H23ClF3N3O5 |
| Synonym | H-AAF-CMK; TPPII Inhibitor |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ64760LB0
|
Thermo Scientific Chemicals
J64760LB0 |
5 mg |
N/A
|
|
|||||
|
AAJ64760MA
|
Thermo Scientific Chemicals
J64760MA |
10 mg |
N/A
|
|
|||||
Description
Tripeptidyl peptidase Inhibitor is a serine protease inhibitor which irreversibly inhibits TPPII (tripeptidyl peptidase II), a giant protease which may substitute for some proteasome functions, and reversibly inhibits Cln (TPPI). AAF-CMK shows only marginal inhibition of the chymotrypsin-like activity of the proteasome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
Chemical Identifiers
| 184901-82-4 | |
| C18H23ClF3N3O5 | |
| H-AAF-CMK; TPPII Inhibitor |
Specifications
| Off-white | |
| A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II | |
| C18H23ClF3N3O5 | |
| H-AAF-CMK; TPPII Inhibitor | |
| Signal Transduction Reagent | |
| ZFZXFWAYCYJFIZ-UHFFFAOYNA-N | |
| 2-amino-N-{1-[(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamoyl]ethyl}propanamide; trifluoroacetic acid |
| -30°C to -10°C | |
| 184901-82-4 | |
| 453.90g/mol | |
| Light Sensitive | |
| Solid | |
| OC(=O)C(F)(F)F.CC(N)C(=O)NC(C)C(=O)NC(CC1=CC=CC=C1)C(=O)CCl | |
| 453.84 |
Safety and Handling
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only