accessibility menu, dialog, popup

UserName

H3CONHH3COHNOClNH2OFFFHO

CAS RN 184901-82-4

IUPAC Name: 2-amino-N-{1-[(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamoyl]ethyl}propanamide; trifluoroacetic acid
Synonyms: 2-amino-N-{1-[(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamoyl]ethyl}propanamide; trifluoroacetic acid
Molecular Weight (g/mol): 453.84
Molecular Formula: C18H23ClF3N3O5
InChi Key: ZFZXFWAYCYJFIZ-UHFFFAOYNA-N
SMILES: OC(=O)C(F)(F)F.CC(N)C(=O)NC(C)C(=O)NC(CC1=CC=CC=C1)C(=O)CCl
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results