missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tripeptidyl Peptidase Inhibitor

Catalog No. AAJ64760LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64760LB0 5 mg
AAJ64760MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64760LB0 Supplier Thermo Scientific Chemicals Supplier No. J64760LB0

Tripeptidyl peptidase Inhibitor is a serine protease inhibitor which irreversibly inhibits TPPII (tripeptidyl peptidase II), a giant protease which may substitute for some proteasome functions, and reversibly inhibits Cln (TPPI). AAF-CMK shows only marginal inhibition of the chymotrypsin-like activity of the proteasome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II

Chemical Identifiers

CAS 184901-82-4
Molecular Formula C18H23ClF3N3O5
Synonym H-AAF-CMK; TPPII Inhibitor

Specifications

Quantity 5 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
Formula Weight 453.90g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid
InChI Key ZFZXFWAYCYJFIZ-UHFFFAOYNA-N
SMILES OC(=O)C(F)(F)F.CC(N)C(=O)NC(C)C(=O)NC(CC1=CC=CC=C1)C(=O)CCl
IUPAC Name 2-amino-N-{1-[(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamoyl]ethyl}propanamide; trifluoroacetic acid
Molecular Weight (g/mol) 453.84
Show More Show Less
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only