missing translation for 'onlineSavingsMsg'
Learn More

Tripelennamine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013235
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013235 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013235 Supplier Merck Emd Millipore Supplier No. T751125G
Only null left

Tripelennamine hydrochloride is an antihistamine drug, which is used in the treatment of bronchitis, asthma and bronchoconstriction.

Specifications

CAS 154-69-8
Physical Form Neat
Linear Formula C16H21N3 · HCl
MDL Number MFCD00012815
Synonym 2-(Benzyl[2-dimethylaminoethyl]amino)pyridine; N-Benzyl-N′,N′-dimethyl-N-(2-pyridyl)ethylenediamine hydrochloride; Pyribenzamine hydrochloride
Type Analytical Standard
Quantity 25 g
Formula Weight 291.82
Grade Analytical Standard