missing translation for 'onlineSavingsMsg'
Learn More

Triolein, tech.

Catalog No. AAJ6241922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6241922 100 g
AAJ6241914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6241922 Supplier Thermo Scientific Chemicals Supplier No. J6241922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 122-32-7 | C57H104O6 | 885.453 g/mol

Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycerol trioleate can be used as an emulsifier, also as creams, lotions and lipsticks matrix. Used as textile finishing agent in textile industry, also used as additive in pigment grind. In pharm industry, used as ointment, emulsifiable paste, suppository, lotion, emulsifier in air agent, stabilizing agent, lubricating agent, antifoaming agents. In food industry, it has the function as emulsify, disperse, defoaming, blister, antistaling of starch and control fat agglutination, usually used as agent in candy, ice cream, cookie and bread. When used in beverage, could avoid ester to float, and avoid protein to sink, therefore to improve stability. When used in bread, cookie , could improve organization structure, increase volume, antistaling, prolong warranty validity.

Solubility
Soluble in chlorofom, ether, and carbon tetrachloride

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C

Chemical Identifiers

CAS 122-32-7
Molecular Formula C57H104O6
Molecular Weight (g/mol) 885.453
MDL Number MFCD00137563
InChI Key PHYFQTYBJUILEZ-BSCDBXJPSA-N
Synonym triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate
PubChem CID 45253964
IUPAC Name [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC

Specifications

Color Yellow
Density 0.91
Boiling Point 235°C to 240°C (18 mmHg)
Flash Point 330°C (626°F)
Refractive Index 1.468
Linear Formula (C17H33COOCH2)2CHOCOC17H33
Quantity 100 g
Beilstein 1718692
Merck Index 14,9732
Solubility Information Soluble in chlorofom, ether, and carbon tetrachloride
Formula Weight 885.45
Percent Purity 75%
Grade Technical
Physical Form Liquid
Chemical Name or Material Triolein
Show More Show Less
EINECSNumber 204-534-7
RTECSNumber RG1936500
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only