missing translation for 'onlineSavingsMsg'
Learn More

Trioctyl Phosphite (mixture) 85.0+%, TCI America™

Catalog No. P038625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P038625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P038625ML Supplier TCI America Supplier No. P038625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3028-88-4
Molecular Formula C24H51O3P
Molecular Weight (g/mol) 418.643
MDL Number MFCD00059943
InChI Key QOQNJVLFFRMJTQ-UHFFFAOYSA-N
Synonym Phosphorous Acid Trioctyl Ester
PubChem CID 76413
IUPAC Name trioctyl phosphite
SMILES CCCCCCCCOP(OCCCCCCCC)OCCCCCCCC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 418.64
Percent Purity ≥85.0% (T)
Physical Form Liquid
Chemical Name or Material Trioctyl Phosphite (mixture)
EINECSNumber (2)-1894
RTECSNumber TH1450000
TSCA No