missing translation for 'onlineSavingsMsg'
Learn More

Trioctadecyl Borate 97.0+%, TCI America™

Catalog No. B052325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B052325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B052325G Supplier TCI America Supplier No. B052325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5337-41-7
Molecular Formula C54H111BO3
Molecular Weight (g/mol) 819.289
MDL Number MFCD00026677
InChI Key GZKLCETYSGSMRA-UHFFFAOYSA-N
Synonym Boric Acid Trioctadecyl Ester, Boric Acid Tristearyl Ester, Tristearyl Borate
PubChem CID 21425
IUPAC Name trioctadecyl borate
SMILES B(OCCCCCCCCCCCCCCCCCC)(OCCCCCCCCCCCCCCCCCC)OCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 343°C
Quantity 25 g
Formula Weight 819.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trioctadecyl Borate
EINECSNumber (2)-2101
RTECSNumber ED5620000
TSCA No