missing translation for 'onlineSavingsMsg'
Learn More

ACCU-SCOPE™ Trinocular with 6MP Excelis HD Camera and Monitor

Catalog No. 01000967
Change view
Click to view available options
Head Style:
Trinocular, 30° Inclined, 360° Rotating
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01000967 Trinocular, 30° Inclined, 360° Rotating
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000967 Supplier ACCU-SCOPE™ Supplier No. EXC360HDS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The new EXC-360-HDS features a Trinocular microscope, with 6MP EXCELIS™ HD camera and monitor w/ 0.35x c-mount.

  • Trinocular viewing head, inclined 30°, rotatable 360°; fixed light path split 50:50
  • Plan Achromat 4x, 10x, 40xR, 100xR oil objectives
  • Excelis HD camera (CAT# AU-600-HDS), 0.35x c-mount
  • PL10x/22mm focusable eyepieces with eye cushions; accepts 24mm reticle
  • Coaxial focusing system with adjustable tension control and upper limit stop to prevent accidental damage to slides
  • Stand: All metal structural frame with molded plastic protective shell; 5v USB-C charging port on back for personal mobile device
  • Stage: Double-layer, rackless mechanical stage with ceramic insert; size 210mm x 170mm; X-Y movement range 78mm x 51mm, Low position right hand controls
  • N.A. 1.25 Abbe Condenser with iris diaphragm; slot for phase contrast or darkfield sliders Coaxial coarse and fine focus controls
  • 3W LED with variable intensity control; LED bulb is rated for over 25,000 hours of continuous use; internal universal power supply 110-240v
  • Accessories: Dust cover, power cord, immersion oil and instruction manual
  • Warranty: 5-year limited warranty for parts, labor and electronic components; 1-year warranty on LED bulb

Specifications

Eyepiece PL-WF 10X/22 mm
Head Style Trinocular, 30° Inclined, 360° Rotating
Lighting Type LED
Magnification Power 40X, 100X, 400X, 1000X
Model ACCU-SCOPE
Objective Types Plan Achromatic
Product Type Digital Microscope System Package
Voltage 110/240 V