missing translation for 'onlineSavingsMsg'
Learn More

Trimyristin 95.0+%, TCI America™

Catalog No. G008825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G008825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G008825G Supplier TCI America Supplier No. G008825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 555-45-3
Molecular Formula C45H86O6
Molecular Weight (g/mol) 723.177
MDL Number MFCD00036229
InChI Key DUXYWXYOBMKGIN-UHFFFAOYSA-N
Synonym trimyristin, glycerol trimyristate, myristin, propane-1,2,3-triyl tritetradecanoate, glyceryl trimyristate, dynasan 114, myristic acid triglyceride, trimyristoylglycerol, tetradecanoic acid, 1,2,3-propanetriyl ester, myristin, tri
PubChem CID 11148
ChEBI CHEBI:77391
IUPAC Name 2,3-di(tetradecanoyloxy)propyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 25 g
Formula Weight 723.18
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trimyristin
EINECSNumber (2)-0669
TSCA Yes