missing translation for 'onlineSavingsMsg'
Learn More

Trimyristin, 90%

Catalog No. AC422090250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422090250 25 g Glass bottle
AC422090050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422090250 Supplier Thermo Scientific Chemicals Supplier No. 422090250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 555-45-3 | C45H86O6 | 722.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 555-45-3
Molecular Formula C45H86O6
Molecular Weight (g/mol) 722.59
MDL Number MFCD00036229
InChI Key DUXYWXYOBMKGIN-UHFFFAOYSA-N
Synonym trimyristin, glycerol trimyristate, myristin, propane-1,2,3-triyl tritetradecanoate, glyceryl trimyristate, dynasan 114, myristic acid triglyceride, trimyristoylglycerol, tetradecanoic acid, 1,2,3-propanetriyl ester, myristin, tri
PubChem CID 11148
ChEBI CHEBI:77391
IUPAC Name 2,3-di(tetradecanoyloxy)propyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 54°C to 58°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9896
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohols,benzene,chloroform,ether
Formula Weight 722.59
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material Trimyristin, 90%
Show More Show Less
EINECSNumber 209-099-7
TSCA TSCA

RUO – Research Use Only