Learn More
FEATHER Trimming Knife Handle
Numéro de catalogue. p-7215437
Change view
Click to view available options
Autoclavable:
Not Autoclavable
Length (Metric):
64.5 mm
80 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Autoclavable Length (Metric)
361021738 Not Autoclavable 80 mm
361027769 Not Autoclavable 64.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 361021738 Fournisseur FEATHER Code fournisseur F80MINI
Il en reste null
FEATHER Trimming Knife Handles attach easily and securely to FEATHER Trimming Knife Blades. Trimming knives are ideal for cutting tissue without causing deformation. Blades sold separately.
  • Made in Japan
  • All Trimming knife handles sold as 1 Pk. except F-80HL and F-80mini which are sold as 5 Pk. and 10 Pk. respectively
Spécifications
Blade Material Plastic
Product Type F-80 MINI; Knife Handle
Length (Metric) 80 mm
For Use With (Equipment) FEATHER Microtome Blade # A35, # A22, # S35, # S22,# N35, # N35HR, # R35, # C35
Description F-80 Mini
Disposable Reusable
Autoclavable Not Autoclavable