missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIML2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5107041

Numéro de catalogue. PIPA5107041


Il en reste null

Description

Description

TRIML2 Polyclonal Antibody for Western Blot

TRIML2 is a member of the tri-partite motif (TRIM) family of proteins. This protein may be regulated by the tumor suppressor p53 and may regulate p53 through the enhancement of p53 SUMOylation. Alternative splicing results in multiple transcript variants.
Spécifications

Identifiants chimiques

TRIML2
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
TRIML2
Probable E3 ubiquitin-protein ligase TRIML2; RING-type E3 ubiquitin transferase TRIML2; SPRY domain-containing protein 6; SPRYD6; TRIML2; tripartite motif family like 2; tripartite motif family-like 2; tripartite motif family-like protein 2
TRIML2
Primary
Affinity Chromatography
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
RUO
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q8N7C3
205860
Synthetic peptide directed towards the middle region of human TRIML2 (aa 215-265).
Antibody

Spécifications

TRIML2
Polyclonal
Unconjugated
TRIML2
Probable E3 ubiquitin-protein ligase TRIML2; RING-type E3 ubiquitin transferase TRIML2; SPRY domain-containing protein 6; SPRYD6; TRIML2; tripartite motif family like 2; tripartite motif family-like 2; tripartite motif family-like protein 2
Rabbit
Affinity Chromatography
RUO
205860
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q8N7C3
TRIML2
Synthetic peptide directed towards the middle region of human TRIML2 (aa 215-265).
100 μL
Primary
Human
Antibody
IgG
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions