missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfoxonium Iodide 98.0+%, TCI America™

Catalog No. T1065500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1065500G 500 g
T106525G 25 g
T1065100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1065500G Supplier TCI America Supplier No. T1065500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1774-47-6
Molecular Formula C3H9IOS
Molecular Weight (g/mol) 220.07
MDL Number MFCD00011899
InChI Key BPLKQGGAXWRFOE-UHFFFAOYSA-M
Synonym trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1
PubChem CID 74498
IUPAC Name trimethyl(oxo)-λ⁶-sulfanylium iodide
SMILES [I-].C[S+](C)(C)=O

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 220.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylsulfoxonium Iodide
RTECSNumber WS3585000
TSCA Yes