missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfoxonium Bromide 98.0+%, TCI America™

Catalog No. T183325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T183325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T183325G Supplier TCI America Supplier No. T183325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25596-24-1
Molecular Formula C3H9BrOS
Molecular Weight (g/mol) 173.068
MDL Number MFCD02093502
InChI Key KEPJZBFFLDRKSF-UHFFFAOYSA-M
PubChem CID 11126750
SMILES C[S+](=O)(C)C.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 173.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylsulfoxonium Bromide