missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium Bromide 98.0+%, TCI America™

Catalog No. T177125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T177125G 25 g
T1771100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T177125G Supplier TCI America Supplier No. T177125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3084-53-5
Molecular Formula C3H9BrS
Molecular Weight (g/mol) 157.069
MDL Number MFCD00051367
InChI Key GOTIICCWNAPLMN-UHFFFAOYSA-M
Synonym trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f
PubChem CID 2735154
IUPAC Name trimethylsulfanium;bromide
SMILES C[S+](C)C.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 157.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylsulfonium Bromide