missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylacetylene 98.0+%, TCI America™

Catalog No. T12395ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12395ML 5 mL
T123925ML 25 mL
T1239250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T12395ML Supplier TCI America Supplier No. T12395ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1066-54-2
Molecular Formula C5H10Si
Molecular Weight (g/mol) 98.22
MDL Number MFCD00008569
InChI Key CWMFRHBXRUITQE-UHFFFAOYSA-N
Synonym trimethylsilylacetylene, trimethylsilyl acetylene, silane, ethynyltrimethyl, ethynyl-trimethyl-silane, ethynyl trimethyl silane, tms acetylene, ethynyltrimethyl silane, trimethylsilyl-acetylene, tmsacetylene
PubChem CID 66111
IUPAC Name ethynyltrimethylsilane
SMILES C[Si](C)(C)C#C

Specifications

Color Colorless
Boiling Point 53°C
Quantity 5 mL
UN Number 1993
Formula Weight 98.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilylacetylene
EINECSNumber (2)-3779
TSCA Yes