missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl cyanide, 97%

Catalog No. AAA1959822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1959822 100 g
AAA1959806 5 g
AAA1959814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1959822 Supplier Thermo Scientific Chemicals Supplier No. A1959822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 7677-24-9 | C4H9NSi | 99.21 g/mol

Trimethylsilyl Cyanide is used as a reagent in the preparation of optically active cyanohydrins. It is used in the derivatization of complex metabolite mixtures by GC-MS. It is employed in the preparation of Reissert compounds, which represent reactive polyamides. Further, it is used to convert pyridine-N-oxides into 2-cyanopyridine. It is also used as a cyanide source in an enantioselective organocatalytic Strecker-type reaction of aliphatic N,N-dialkylhydrazones. In addition to this, it is used as a reagent for the cyanosilylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl Cyanide is used as a reagent in the preparation of optically active cyanohydrins. It is used in the derivatization of complex metabolite mixtures by GC-MS. It is employed in the preparation of Reissert compounds, which represent reactive polyamides. Further, it is used to convert pyridine-N-oxides into 2-cyanopyridine. It is also used as a cyanide source in an enantioselective organocatalytic Strecker-type reaction of aliphatic N,N-dialkylhydrazones. In addition to this, it is used as a reagent for the cyanosilylation of aldehydes.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Store in a cool place.Incompatible with water, strong oxidizing agents, strong reducing agents, strong acids and strong bases.

Chemical Identifiers

CAS 7677-24-9
Molecular Formula C4H9NSi
Molecular Weight (g/mol) 99.21
MDL Number MFCD00001765
InChI Key LEIMLDGFXIOXMT-UHFFFAOYSA-N
Synonym trimethylsilyl cyanide, cyanotrimethylsilane, trimethylsilanecarbonitrile, trimethylsilylcyanide, silanecarbonitrile, trimethyl, trimethylsilylnitrile, tmscn, trimethyl silane cyanide, trimethylsilylcarbonitrile, tms cyanide
PubChem CID 82115
IUPAC Name trimethylsilylformonitrile
SMILES C[Si](C)(C)C#N

Specifications

Boiling Point 118°C to 119°C
Melting Point 11°C to 12°C
Quantity 100 g
Flash Point 1°C (33°F)
Odor Almond-like
Refractive Index 1.391
Sensitivity Moisture sensitive
Density 0.783
UN Number UN3383
Beilstein 1737612
Solubility Information Miscible with organic solvents.
Formula Weight 99.21
Percent Purity 97%
Chemical Name or Material Trimethylsilyl cyanide
Show More Show Less
Hazard Category H225-H300+H310+H330
Hazard Statement GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapour.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
EINECSNumber 231-657-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only