missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene Sulfide 98.0+%, TCI America™

Catalog No. T04745ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04745ML 5 mL
T047425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04745ML Supplier TCI America Supplier No. T04745ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 287-27-4
Molecular Formula C3H6S
Molecular Weight (g/mol) 74.14
MDL Number MFCD00042751
InChI Key XSROQCDVUIHRSI-UHFFFAOYSA-N
Synonym trimethylene sulfide, thiacyclobutane, propane, 1,3-epithio, unii-1o7c19qqdb, 1o7c19qqdb, 1,3-epithio propane, acmc-209h4c, trimethylene sulfide 8ci, trimethylene sulfide gc, inchi=1/c3h6s/c1-2-4-3-1/h1-3h
PubChem CID 9251
ChEBI CHEBI:87565
IUPAC Name thietane
SMILES C1CSC1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 mL
UN Number 1993
Formula Weight 74.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylene Sulfide