missing translation for 'onlineSavingsMsg'
Learn More

Trimethylene Bis(4-aminobenzoate) 98.0+%, TCI America™

Catalog No. T164625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T164625G 25 g
T1646100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T164625G Supplier TCI America Supplier No. T164625G
Only null left

Chemical Identifiers

CAS 57609-64-0
Molecular Formula C17H18N2O4
Molecular Weight (g/mol) 314.34
MDL Number MFCD00129765
InChI Key YPACMOORZSDQDQ-UHFFFAOYSA-N
Synonym 1,3-Bis(4-aminobenzoyloxy)propane, 1,3-Propanediol Bis(4-aminobenzoate)
PubChem CID 93312
IUPAC Name 3-(4-aminobenzoyloxy)propyl 4-aminobenzoate
SMILES NC1=CC=C(C=C1)C(=O)OCCCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 128°C
Quantity 25 g
Formula Weight 314.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylene Bis(4-aminobenzoate)
EINECSNumber (3)-3715
TSCA Yes