missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine hydrochloride, 98%

Catalog No. AAL0471830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0471830 250 g
AAL0471818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0471830 Supplier Thermo Scientific Chemicals Supplier No. L0471830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 593-81-7 | C3H10ClN | 95.57 g/mol

Trimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.

Solubility
Soluble in water (737 mg/ml at 20°C), chloroform (moderately), and alcohol. Insoluble in ether.

Notes
Hygroscopic. Store under dry inert gas and away from moisture. Incompatible with oxidizing agents and water.

Chemical Identifiers

CAS 593-81-7
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012478
InChI Key SZYJELPVAFJOGJ-UHFFFAOYSA-N
Synonym trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride
PubChem CID 10313079
ChEBI CHEBI:64700
IUPAC Name N,N-dimethylmethanamine;hydrochloride
SMILES [H+].[Cl-].CN(C)C

Specifications

Melting Point ∽280°C (decomposition)
Quantity 250 g
Beilstein 3905063
Sensitivity Hygroscopic
Merck Index 14,9710
Solubility Information Soluble in water (737mg/ml at 20°C),chloroform (moderately),and alcohol. Insoluble in ether.
Formula Weight 95.57
Percent Purity 98%
Chemical Name or Material Trimethylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-810-0
RTECSNumber YH2700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only