missing translation for 'onlineSavingsMsg'
Learn More

Triméthylamine (environ 28% en eau, environ 4,3 mol/L), TCI America™

Numéro de catalogue. T0464500ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T0464500ML 500 mL
T046425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T0464500ML Fournisseur TCI America Code fournisseur T0464500ML

Identifiants chimiques

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
IUPAC Name Triméthylamine
SMILES CN(C)C

Spécifications

Color Incolore
Quantity 500 mL
UN Number 1297
Formula Weight 59.11
Physical Form Liquide
Chemical Name or Material Trimethylamine (ca. 28% in Water, ca. 4.3mol/L)
EINECSNumber (2)-0140
RTECSNumber PA0350000
TSCA Yes
Recommended Storage Réfrigérateur